missing translation for 'onlineSavingsMsg'
Learn More

Congo Red MP Biomedicals

Product Code. 11491842 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
Product Code. Quantity unitSize
11491842 25 g 25g
1 options
This item is not returnable. View return policy

Product Code. 11491842

Brand: MP Biomedicals 0210509925

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 573-58-0
Molecular Formula C32H22N6Na2O6S2
Synonym Direct Red 28
InChI Key IQFVPQOLBLOTPF-UHFFFAOYSA-L
SMILES C1=CC=C2C(=C1)C(=CC(=C2N)N=NC3=CC=C(C=C3)C4=CC=C(C=C4)N=NC5=C(C6=CC=CC=C6C(=C5)S(=O)(=O)[O-])N)S(=O)(=O)[O-].[Na+].[Na+]
IUPAC Name disodium;4-amino-3-[[4-[4-[(1-amino-4-sulfonatonaphthalen-2-yl)diazenyl]phenyl]phenyl]diazenyl]naphthalene-1-sulfonate
Molecular Weight (g/mol) 696.664
PubChem CID 11313
ChEBI CHEBI:34653
Formula Weight 696.7
Quantity 25 g
Chemical Name or Material Congo Red
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.