Learn More
BRAND™ Conical Joint Stopper
Product Code. 10578211 Shop All Brand Products
Change view
Click to view available options
Standard Taper Joint:
NS 10/19
NS 34/35
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Standard Taper Joint Unit Size
10578211 NS 34/35 Each
10653012 NS 10/19 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10578211 Supplier BRAND™ Supplier No. 144640
  • ONLINE EXCLUSIVE PRICE
    17.00 €
    16.15 € / Pack of 1
    Save 0.85 €
    5% Off
Estimated Shipment: 08-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Conical Joint Stopper features a hollow design and hexagonal grip.
  • Borosilicate Glass 3.3
  • DIN 12252
  • Hollow with hexagonal grip
TRUSTED_SUSTAINABILITY
Specifications
Description Hollow with hexagonal grip
Standard Taper Joint NS 34/35
Material Borosilicate Glass
Shape Conical