missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Conivaptan hydrochloride, MedChemExpress
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
Description
Conivaptan (hydrochloride) is a non-peptide antagonist of vasopressin receptor, with Ki values of 0.48 and 3.04 nM for rat liver V1A receptor and rat kidney V2 receptor respectively.
Specifications
Specifications
| Chemical Name or Material | Conivaptan hydrochloride |
| CAS | 168626-94-6 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C32H27ClN4O2 |
| Quantity | 50 mg |
| Synonym | YM 087 |
| Solubility Information | DMSO : ≥ 100 mg/mL (186.90 mM) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble) |
| SMILES | O=C(C1=CC=CC=C1C2=CC=CC=C2)NC3=CC=C(C(N4CCC(N=C(C)N5)=C5C6=CC=CC=C64)=O)C=C3.Cl |
| Molecular Weight (g/mol) | 535.04 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction