Learn More
Emma Safety Footwear Constans Safety Shoes
Product Code. 15998211
Change view
Click to view available options
Size:
39
41
42
43
44
45
47
Unit Size:
1 pair
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
15998211 39 1 pair
15908221 41 1 pair
15918221 42 1 pair
15928221 43 1 pair
15938221 44 1 pair
15948221 45 1 pair
15958221 47 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 15998211 Supplier Emma Safety Footwear Supplier No. MM13207039
  • ONLINE EXCLUSIVE PRICE
    424.20 €
    266.70 € / 1 pairs
    Save 157.50 €
    37% Off
Estimated Shipment: 15-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High quality full grain leather upper and nappa leather lining. Exclusively developed for semi-orthopedic adjustments especially where customized in-soles are required.
  • S3-Safety shoe with sole plate for wet work
  • Full grain leather construction
  • Vibram™ Nitrile Rubber Heat Resistant Outsole
Specifications
Certifications/Compliance EN ISO 20345:2011
Closure Type Laces
Color Black
Design Constans
Liner Material Full grain Nappa leather
Material Leather
Size 39
Toe Material Aluminum