missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Consumables and Services Kits for Orion™ 2110XP Ammonia Analyzer

Product Code. 17128988
Change view
Click to view available options
Type:
Ammonia Consumables Kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
17128988 Ammonia Consumables Kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17128988 Supplier Thermo Scientific™ Supplier No. 211050XP
  • ONLINE EXCLUSIVE PRICE
    3522.53 €
    2948.17 € / Pack of 1
    Save 574.36 €
    16% Off
 Restricted Item
Estimated Shipment: 15-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Keep needed supplies and service items in stock to avoid lost productivity with Thermo Scientific™ Consumables and Services Kits for Orion™ 2110XP Ammonia Analyzer.

Keep needed supplies and service items in stock to avoid lost productivity with Thermo Scientific™ Consumables and Services Kits for the Thermo Scientific™ Orion™ 2110XP Ammonia Analyzer.

TRUSTED_SUSTAINABILITY

Specifications

Accuracy (mV) ±(0.5 mV + 0.1%)
Accuracy (Temperature) ±0.5 °C
Description Ammonia Consumables Kit for one year operation
Temperature (Metric) Operating -10°C to +120°C
Type Ammonia Consumables Kit
Display Type 211050XP
Flow Rate 40 mL/min.
Includes 2 x 2L acetic acid reagent (181030), 2 x 4 ft. diffusion tubing assemblies with O-rings (150060), reagent bottle adapter (2110RBA), ammonia calibration standards kit (181040), ammonia electrode (100047), reference electrode (210056), 5 x 2 oz. reference electrode filling solution (181073), O-ring kit (2110OK), restriction tube assembly (2100RT), 4 inlet filters (181120) and box of pipette tips (204846-001)
Inlet Pressure 8 to 100 psig
Inputs 85-132; 170-264 VAC
Outputs Alarm: 3; Relay: 3; Analog: 2; Selections: 0-20mA; 4-20 mA
Range (mV) ±1999.9 m
Relay Details Max Load: 250VAC; 30VDC; 5A
Resolution (mV) 0.1 mV
Resolution (Temperature) 0.1 °C
Response Time 211050XP
Sample Inlet 0.25 NPTF Tube Fitting in.
Sample Temperature 5 to 45 °C
Show More Show Less

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.