missing translation for 'onlineSavingsMsg'
Learn More

Manutan Containers, Euro size, blue, 125mm x 150mm x 200mm

Product Code. 12894578
Encompass_Preferred
This item is not returnable. View return policy

Product Code. 12894578

Brand: Manutan 109B061N

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Multi-purpose plastic bin range made from robust polypropylene. Euro sized to stack on standard Euro pallets 1200mm x 800mm and 1200mm x 1000mm. Heavy load capacity when stacked due to corner design. Slide down transparent vision panel allows bins to used either as full height containers with 100% cube utilisation or as semi-open fronted bins. All models include large plain label card. Can be used on louvred panels (except 18. 50ltr container). Flat bottom on all models assists use on conveyors. Suitable for use to -20°C.
  • Type B
  • Blue
  • Dimensions (H x W x D): 125mm x 150mm x 200mm
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.