missing translation for 'onlineSavingsMsg'
Learn More

Contec™ EasyReach™ Cleaning Pads

Contec™'s EasyReach™ cleaning pads are very absorbent, which ensures even coverage of disinfectant or IPA.

Brand:  Contec™ MEQT0021

342.00 EUR valid until 2024-12-31
Use promo code "22912" to get your promotional price.



Additional Details : Weight : 3.10000kg

 View more versions of this product

Product Code. 16691872

  • 459.00 € / Pack of 100
Estimated Shipment: 02-07-2024
to see stock.
Add to basket

Description

Description

  • Single-use sealed polyester
  • Provide a low level of particulates which is essential for use in product contact areas
  • Very absorbent, ensuring good even coverage of disinfectant or IPA
  • Easy grab edge on the cleaning pad ensures the pad fits tightly on the mop frame
  • Sterile cleaning pads are presaturated with 70% IPA
Specifications

Specifications

7.5 x 2.8 in. (19 x 7 cm)
2.8 in.
7 cm
19 cm
Yes
4/Bag; 25 Bags/Cs.
Cleaning Pads
Non-sterile
Not Autoclavable
7.5 in.
4 Ea./Bag, 25 Bags/Case
rigid, flexible film, half suits (CAI, CACI), Restricted Access Barrier Systems (RABS), Filling Lines, Laminar Air Flow Hoods (LAF), Bio-Safety Cabinets (BSC), Glove Boxes, Incubators
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers