missing translation for 'onlineSavingsMsg'
Learn More

Contec™ HydroPure
BIOPHARMA

Product Code. 15686986 Shop All Contec Products
Change view
Click to view available options
Quantity:
2 x 5 L
6 x 1 L
Container Type:
Capped Bottle
Trigger Spray Bottle
Unit Size:
Pack of 2
Pack of 6
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Container Type Unit Size
15686986 6 x 1 L Trigger Spray Bottle Pack of 6
15676986 2 x 5 L Capped Bottle Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15686986 Supplier Contec™ Supplier No. FBT16HP
  • ONLINE EXCLUSIVE PRICE
    114.00 €
    91.05 € / Pack of 6
    Save 22.95 €
    20% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Blend of 6% hydrogen peroxide and purified water, low residue, sporicidal biocide. HydroPure is filtered to 0.2μ leaves no residue, eliminating any residue removal stage.

Blend of 6% hydrogen peroxide and purified water, low residue, sporicidal biocide. Contec™ HydroPure is filtered to 0.2μ leaves no residue, eliminating any residue removal stage.

  • Efficacious against bacteria, fungi, moulds, yeasts and spores
  • 0.2μm filtered and filled under Grade A uni-directional flow air and bagged in Grade B cleanroom
  • Leaves little to no residue and has endotoxin level of <0.25 EU/mL
  • Safe to use in product contact areas
  • Designed for ease of entry into pharmaceutical cleanrooms
  • Ensures the product is free from contamination and particulates
  • Sufficient sporicidal activity for Grade A/B area
  • Large droplet size reduces the risk of inhalation and provides good surface coverage
  • Bagged in polyethylene linear tear bags
  • Facilitates transfer disinfection into isolators and RABS
  • RTU with one year shelf life
TRUSTED_SUSTAINABILITY

Specifications

Product Type Hydrogen Peroxide and Purified Water Blend
Form Liquid
pH Not Applicable
Scent Unscented
Sterility Non-sterile
Appearance Clear
Autoclavable Not Autoclavable
Color Clear
Composition 6% hydrogen peroxide in purified water
Container Type Trigger Spray Bottle
Disposable Limited-use
Filtered Filtered
Flammable Yes
Quantity 6 x 1 L
Volume (Metric) 1 L
Show More Show Less
compliance-icons
Product Identifier
  • contec hydropure
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.