missing translation for 'onlineSavingsMsg'
Learn More

Contec™ PROSAT™ Sterile Presaturated Nonwoven Wipes
GREENER_CHOICE

Product Code. 11932435 Shop All Contec Products
Change view
Click to view available options
Dimensions (L x W):
28 x 23 cm (11 x 9 in.)
Unit Size:
Pack of 30
Product Code. Dimensions (L x W) unitSize
11932435 28 x 23 cm (11 x 9 in.) Pack of 30
1 options
This item is not returnable. View return policy

Product Code. 11932435

Brand: Contec™ PS911EB/ETOH

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Wipe down isolators, cabinets, hoods, and other cleanroom equipment and tools. Contec™ PROSAT™ Sterile Presaturated Nonwoven Wipes are ideal for cleanup following BD RODAC™ plate sampling. These wipes are designed specifically for sterile environments.

  • Designed for pharmaceutical, biotech, and medical device applications
  • Choose spunlaced nonwoven 100% polyester wipes for improved cleanliness, economical hydroentangled polyester/cellulose wipers, or meltblown polypropylene wipes for improved durability
  • Conveniently presaturated with 70/30 blend of semi-conductor grade IPA and DI water or 70/30 blend of denaturated ethyl alcohol and DI water to reduce VOC emissions and hazardous waste
  • All wipes are sterilized by gamma radiation to a 10-6 Sterility Assurance Level and validated per AAMI ISO 11137 (Method 1) guidelines
  • Certificate of Sterility and Certificate of Irradiation included
  • Recommended for use in ISO Class 5 (FED-STD-209E Class 100/M3.5) environments
  • Individually lot numbered resealable pouches with tamper-evident labels ensure product integrity and cleanliness
TRUSTED_SUSTAINABILITY

Specifications

Product Type Pre-saturated Wipe
Length (English) 11 in.
Length (Metric) 28 cm
Width (English) 11 in.
Width (Metric) 23 cm
Material Polypropylene
Autoclavable Not Autoclavable
Certifications/Compliance ISO Class 4
Disposable Single-use
Dimensions (L x W) 28 x 23 cm (11 x 9 in.)
For Use With (Application) Ideal for routine cleaning of tools, instruments and other laboratory equipment; designed for sterile environments
Packaging Quantity 30 Wipes/Bag
Ply 1-ply
Saturation Solution 70% Denatured Ethyl Alcohol/30% DI Water
Sterility Sterile
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • CONTEC PROSAT STERILE MBPP WITH 70% DENATURED ETHANOL
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P370+P378-In case of fire: Use dry chemical/carbondioxide to extinguish.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P411-Store at temperatures not exceeding 40°C/104°F.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : ETHANOL, PROPAN-2-OL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.