Learn More
Memmert™ Convection Oven Stainless Steel Grids
Product Code. 12607057 Shop All Memmert Products
Change view
Click to view available options
For Use With (Equipment):
Model 30
Models 110, 160 incubator
Models 260 and 410
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
12607057 Model 30 Each
12637057 Models 260 and 410 Each
11711464 Models 110, 160 incubator Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12607057 Supplier Memmert™ Supplier No. E28884
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Use these grids with Memmert ovens. Memmert™ Convection Oven Stainless Steel Grids feature sturdy construction and are available in a variety of sizes to fit your needs.
  • Reinforced stainless steel grid
  • For convection ovens
Specifications
For Use With (Equipment) Model 30
Type Grid
Material Stainless Steel