missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ Protective Coverall 4532+, Cool White
Description
- Unique liquid repellency with oil and alcohol resistance material treatment
- Soft and strong multi-layered 52gsm SSMMS for strong performance
- Made from super breathable SSMMS non-woven for effective release of body heat and moisture and reduced heat stress
- 3 panel hood, reinforced triangular gusset panel, 2-way zip and storm flap with sealable tape, elasticated back and ankles to better freedom of movement
- Anti-static coating EN 1149-5 reduces static discharge and the risk of sparks
- EN 1073-2 nuclear particulate hold out
- Knitted cuffs and elastic waist and ankles for a better fit and freedom of movement
Specifications
Specifications
| Product Type | Protective Coveralls |
| Size | Medium |
| Gender | Unisex |
| Material | SSMMS |
| Color | White |
| Disposable | Single-use |
| Waist Style | Elastic |
| Cuff Style | Knit |
| Closure Type | Front Zipper |
| Sterility | Non-sterile |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?