Learn More
Corning™ Extension Collar for CoolBox™ XT System
For use with CoolBox™ XT System. Corning™ Extension Collar is required when using taller CoolRack™ tube modules and tall tubes in CoolBox system.
Brand:  Corning™ 432086
Code : NEW
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 15512861
  • 102.00 € / Each
Add to basket
Alert:
Lid closure not possible with addition of extension collar
Description
Description
For use with CoolBox XT and CoolBox 2XT to extend the height when using tall tube modules. The collar is magnetized and easily adheres to the unit base.
Specifications
Specifications
Pink
Extension Collar
CoolBox XT and CoolBox 2XT
20.0 x 16.0 x 7.0 cm (7.8 x 6.2 x 2.7 in.) Interior: 14.0 x 9.4 x 7.0 cm (5.5 x 3.7 x 2.7 in.)
20 cm
16 cm
7 cm
Square
Extend the Height When Using Tall Tube Modules
1/Pk.
7.8 in.
6.2 in.
2.75 in.