missing translation for 'onlineSavingsMsg'
Learn More

Corning™ CoolCell™ LX Cell Freezing Vial Containers
BIOPHARMA

Product Code. 15813885
Change view
Click to view available options
Color:
Assorted (Green, Orange, Pink, Purple)
Green
Orange
Pink
Purple
Quantity:
1/Pk., 1/Cs.
4/Pk., 1/Cs.
Unit Size:
Each
Pack of 4
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Quantity Unit Size
15813885 Assorted (Green, Orange, Pink, Purple) 4/Pk., 1/Cs. Pack of 4
15552771 Purple 1/Pk., 1/Cs. Each
15562771 Green 1/Pk., 1/Cs. Each
15572771 Orange 1/Pk., 1/Cs. Each
15582771 Pink 1/Pk., 1/Cs. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 15813885 Supplier Corning™ Supplier No. 432138
  • ONLINE EXCLUSIVE PRICE
    725.00 €
    691.48 € / Pack of 4
    Save 33.52 €
    4% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Provides alcohol-free freezing at the rate of -1°C/minute when placed in a -80°C freezer.

Corning CoolCell, in combination with a -80°C freezer, will provide alcohol-free freezing at the rate of -1°C/minute that is ideal for cryopreservation of most cells and cell lines. Using a combination of uniform-density cross-linked polyethylene foam, a solid state core, and radial vial symmetry, freezing profiles are consistent and reproducible. The foam is non-absorbent and will impose negligible change in the freezer environment; thereby protecting nearby frozen samples. The removal of vials is accomplished by tipping the device upside down so the vials slide out.

Features: Unique features of controlled-rate freezing with Corning™ CoolCell™ include:

  • Ease of use
  • Alcohol and fluid-free freezing
  • Lower cost of use than alcohol-based devices
  • High cell recovery and cell viability
  • Reproducibility
  • Simple, consistent way to standardize controlled-rate freezing
TRUSTED_SUSTAINABILITY

Specifications

Color Assorted (Green, Orange, Pink, Purple)
Material Polyethylene Foam
Shape Round
Type Freezer Box
For Use With (Application) Cryopreservation of Most Cells and Cell Lines
Holds 12 (Standard 1 to 2 mL Cryogenic Vials)
Quantity 4/Pk., 1/Cs.
Dimensions (L x W) 11.7 x 10.11 cm
Length (Metric) 11.7 cm
Width (Metric) 10.11 cm
Diameter (Metric) Well 12.7 mm
Weight (Metric) 0.5 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.