missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ CoolRack™ SV Sample Vial Modules

Click to view available options
Holds:
12 x 10 mL sample vials
12 x 5 mL sample vials
Design:
SV10
SV2
Unit Size:
Each
Description
Corning CoolRack thermo-conductive tube modules eliminate variability which originates from tubes placed directly into ice, dry ice, alcohol baths, water baths and other temperature sources. Place the CoolRack module directly onto a temperature source between -196°C to > 100°C and it will rapidly adapt to that temperature.
- Cell therapy injectable ampule modules
- Eensures ± 0.1°C temperature uniformity across all tubes when cooling, snap freezing, heating or thawing.
- All CoolRack modules may be autoclaved, high heat sterilized or decontaminated with bleach, alcohol or other disinfectants or lab detergents.
- Suggested applications include cooling reagents such as restriction enzymes, dNTPs and antibodies, alcohol-free dry ice snap freezing of tissue, virus and bacteria samples and bench top cryogenic tube sorting in liquid nitrogen.
- Certain CoolRack modules are SBS-compatible.
Specifications
Specifications
| Holds | 12 x 10 mL sample vials |
| Design | SV10 |
| Type | SV Sample Vial Module |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction