missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Coomassie Blue R250, for Electrophoresis, (Page Blue 83) C.I. 42660, Fisher Chemical™
Shop All Fisher Chemical ProductsDescription
For use in staining proteins in SDS-PAGE gels.
Specifications
Specifications
| CAS | 6104-59-2 |
| Color | Dark Blue |
| pH Range | 6.2 |
| Form | Powder Solid |
| Melting Temperature | 174°C/345.2°F |
| Molecular Weight (g/mol) | 825.96g/mol |
| Quantity | 25 g |
| Product Type | Protein Stain |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?