missing translation for 'onlineSavingsMsg'
Learn More

Coomassie Brilliant Blue R-250, Fisher BioReagents

Product Code. 10573165
Click to view available options
Quantity:
25 g
50 g
Packaging:
Amber Glass
Glass Bottle
missing translation for 'unitSize'
25g
50g
This item is not returnable. View return policy

Product Code. 10573165

missing translation for 'mfr': Fisher BioReagents BP10150

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Brilliant Blue R-250 is commonly used for staining proteins in polyacrylamide gels.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6104-59-2
Molecular Formula C45H44N3NaO7S2
Molecular Weight (g/mol) 825.97
MDL Number MFCD00041762
InChI Key HAEGGLCXLFWTBE-UHFFFAOYSA-M
Synonym Coomassie Blue R-250
PubChem CID 61365
IUPAC Name sodium;3-[[4-[[4-(4-ethoxyanilino)phenyl]-[4-[ethyl-[(3-sulfonatophenyl)methyl]azaniumylidene]cyclohexa-2,5-dien-1-ylidene]methyl]-N-ethylanilino]methyl]benzenesulfonate
SMILES [Na+].CCOC1=CC=C(NC2=CC=C(C=C2)[C+](C2=CC=C(C=C2)N(CC)CC2=CC=CC(=C2)S([O-])(=O)=O)C2=CC=C(C=C2)N(CC)CC2=CC=CC(=C2)S([O-])(=O)=O)C=C1

Specifications

Melting Point 174°C
Extinction Coefficient 700g(-1) cm(-1) min.
Identification Pass Test
Lambda Max 590 to 594nm
Formula Weight 825.96
Loss on Drying 5% max.
Packaging Amber Glass
Density 0.25g/cm³
Color Index No. 42660
Color Brown/Red
pH 6.2
Quantity 50 g
Physical Form Powder
Chemical Name or Material Brilliant Blue R-250
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.