missing translation for 'onlineSavingsMsg'
Learn More

New Pig™ Copolymer Polypropylene Overpack Salvage Drum

Product Code. 12961405
Encompass_Preferred
Change view
Click to view available options
Capacity (Metric):
76 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
12961405 76 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12961405 Supplier New Pig™ Supplier No. PAK708WH
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Product Type Overpack Salvage Drum
Capacity (Metric) 76 L
Color White
For Use With (Application) Ideal for regulated packaging applications, including shipping hazardous waste by land, sea or rail
Material PPC
Height (Metric) 48 cm
Weight (Metric) 5.89 Kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.