missing translation for 'onlineSavingsMsg'
Learn More

Copovidone (Technical Grade), TRC

Product Code. 30351734
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30351734 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30351734 Supplier Toronto Research Chemicals Supplier No. TRCC6852401G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Copovidone
CAS 25086-89-9
Molecular Formula C6H9NO.C4H6O2
Quantity 1 g
Synonym Acetic Acid Vinyl Ester Polymer with 1-Vinyl-2-pyrrolidinone, 1-Ethenyl-2-pyrrolidinone Polymer with Ethenyl Acetate, 1-Vinyl-2-pyrrolidinone Polymer with Vinyl Acetate, 1-Vinyl-2-pyrrolidone-vinyl Acetate Copolymer, 1-Vinylpyrrolidone-vinyl Acetate Copolymer, Agrimer VA 03E, Agrimer VA 05E, Agrimer VA 07E, Agrimer VA 3, Agrimer VA 31, Agrimer VA 3E, Agrimer VA 5, Agrimer VA 5I, Agrimer VA 6, Agrimer VA 64, Agrimer VA 7W, Akon KS, Akorn KS, Copolyvidon, Copolyvidonum, Copovidon, Copovidone, Copovidone VA 64, E 335, E 335 (polymer), E 535, E 635, E 635 (polymer), E 735, Eukaline 480, GAF-E 735, GAF-S 630, Ganex E 535, Gantron PVP, Gantron S 630, Gantron S 860, I 335, I 535, I 635, I 735, Kolima 10, Kolima 35, Kolima 75, Kollidon VA 64, Luviscol VA 55I, Luviskol 64, N-Vinyl-2-pyrrolidone-vinyl Acetate Copolymer, N-Vinylpyrrolidin-2-one-vinyl Acetate Copolymer, N-Vinylpyrrolidone-vinyl Acetate Copolymer, N-Vinylpyrrolidone-vinyl Acetate Copolymer, N-Vinylpyrrolidone-vinyl Acetate Polymer, NSC 114023, NSC 114024, NSC 114025, NSC 114026, P 0382, P 0385, PVA 6450, PVP-VA, PVP-VA 735, PVP-VA-E 735, PVP-VA-W 735, PVP/VA 630, PVP/VA E 335, PVP/VA E 535, PVP/VA-S 630, PVP/VA-S 630L, PVP/Va copolymer, PVPNA-S 630, PVPVA 64, PVPVA 7/30, Plasdone S 630, Pluronic S 630, Polectron 845, Poly(N-vinylpyrrolidone-vinyl Acetate), Poly(vinyl Acetate-co-vinyl pyrrolidone), Poly(vinyl Acetate-vinylpyrrolidinone), Poly(vinylpyrrolidone-vinyl Acetate), R 219, R 219 (Vinyl Polymer), S 630, VA 46E, VA 64, Vinyl acetate-1-vinyl-2-pyrrolidinone Copolymer, Vinyl Acetate-1-vinyl-2-pyrrolidinone Polymer, Vinyl Acetate-1-vinyl-2-pyrrolidone Copolymer, Vinyl acetate-N-vinyl-2-pyrrolidinone copolymer, Vinyl Acetate-N-vinyl-2-pyrrolidone Copolymer, r, W 635, W 635 (vinyl polymer), W 735
InChI Formula InChI=1S/C6H9NO.C4H6O2/c1-2-7-5-3-4-6(7)8;1-3-6-4(2)5/h2H,1,3-5H2;3H,1H2,2H3
SMILES C=CN(CCC1)C1=O.C=COC(C)=O
IUPAC Name 1-vinylpyrrolidin-2-one vinyl acetate
Molecular Weight (g/mol) 197.23
Formula Weight 197.11
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.