missing translation for 'onlineSavingsMsg'
Learn More

Copper, 99.9998%, (trace metal basis), Thermo Scientific™

Product Code. 10795842 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
125 g
Packaging:
Glass bottle
Unit Size:
125g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10795842 125 g Glass bottle 125g
10555090 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10795842 Supplier Thermo Scientific Acros Supplier No. 437051250
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

For ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-50-8
Molecular Formula Cu
Molecular Weight (g/mol) 63.55
MDL Number MFCD00010965
InChI Key RYGMFSIKBFXOCR-UHFFFAOYSA-N
Synonym cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
PubChem CID 23978
ChEBI CHEBI:30052
IUPAC Name copper
SMILES [Cu]

Specifications

Color Brown-Rust
Melting Point 1083.0°C
Boiling Point 2595.0°C
Quantity 125 g
Packaging Glass bottle
Assay Percent Range 99.9998%
Form Shot
Linear Formula Cu
Solubility Information Solubility in water: insoluble.
Formula Weight 63.54
Percent Purity 99.9998%
Grade Trace Metal Basis
Chemical Name or Material Copper
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.