missing translation for 'onlineSavingsMsg'
Learn More

Copper(I) Chloride, ≥97.0% (RT), Honeywell Fluka™

Product Code. 15656020 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
25 g
250 g
1 kg
Packaging:
Poly bottle
Unit Size:
1kg
250g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15656020 25 g Poly bottle 25g
15646020 250 g Poly bottle 250g
15636020 1 kg Poly bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15656020 Supplier Honeywell-Fluka Supplier No. 6116825G
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Puriss. p.a., ACS Reagent,≥97.0% (RT)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7758-89-6
Molecular Formula BrCu
Molecular Weight (g/mol) 143.45
MDL Number MFCD00010969
InChI Key NKNDPYCGAZPOFS-UHFFFAOYSA-M
Synonym copper i bromide, copper bromide cubr, copper 1+ bromide, copper monobromide, cuprousbromide, hsdb 270, cubr, copper 1 bromide
PubChem CID 24593
IUPAC Name λ¹-copper(1+) chloride
SMILES [Cu+].[Br-]

Specifications

Boiling Point 1345°C (1013hPa)
Melting Point 492°C
Color Colorless
Physical Form Powder
Quantity 25 g
Linear Formula CuBr
UN Number NONH for all modes of transport
Formula Weight 143.45g/mol
Percent Purity ≥98.0% (RT)
Odor Odorless
Packaging Poly bottle
Density 4.98g/cm3
Chemical Name or Material Copper(I) bromide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.