missing translation for 'onlineSavingsMsg'
Learn More

Copper(I) chloride, 99%, extra pure, purified

Product Code. 10561841 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10561841 100 g Glass bottle 100g
10338200 25 g Glass bottle 25g
15199223 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10561841 Supplier Thermo Scientific Acros Supplier No. 208391000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7758-89-6 | ClCu | 99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7758-89-6
Molecular Formula ClCu
Molecular Weight (g/mol) 99
MDL Number MFCD00010971
InChI Key OXBLHERUFWYNTN-UHFFFAOYSA-M
Synonym cuprous chloride, copper i chloride, copper chloride cucl, copper monochloride, dicopper dichloride, copper 1+ chloride, cucl, chlorid medny czech
PubChem CID 62652
ChEBI CHEBI:53472
IUPAC Name chlorocopper
SMILES Cl[Cu]

Specifications

Boiling Point 1490.0°C
Melting Point 430.0°C
Color Gray-Green
Physical Form Crystalline Powder
Quantity 100 g
Assay Percent Range 98.5 to 101.5% (Complexometry)
Linear Formula CuCl
Fieser 01,166; 02,91; 03,67; 04,109; 05,164; 06,145; 07,80; 08,118; 09,123; 11,140; 12,141; 13,152; 14,100; 15,101; 16,97
Merck Index 15,265
Solubility Information Solubility in water: 0.06g/L (25°C). Other solubilities: soluble in conc. hcl and nh4oh, forming complexes, practically insoluble in alcohol and acetone
Formula Weight 99
Percent Purity 99%
Grade Extra Pure
Packaging Glass bottle
Specific Gravity 4.14
Density 4.1400g/mL
Chemical Name or Material Copper(I) chloride, 99%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Copper(I) chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H315-Causes skin irritation.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.