missing translation for 'onlineSavingsMsg'
Learn More

Copper(I) chloride, 99% (metals basis)

Product Code. 11394797
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11394797 500 g 500g
11384797 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11394797 Supplier Thermo Scientific Alfa Aesar Supplier No. 041837.36
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7758-89-6 | ClCu | 98.996 g/mol

Copper(I) chloride is precursor to many copper compounds including copper oxychloride and many organocuprate compounds of synthetic interest. It is used as a catalyst as well as a reagent in many organic reactions including Gatterman-Koch, Sandmeyer, Grignard and Gilman reactions. It catalyzes 1,4-addition of Grignard reagents to alpha,beta-unsaturated ketones. It is also used as a doping salt to enhance the cationization of polystyrene in matrix-assisted laser desorption/ionization mass spectrometry. It is also an intermediate formed in the Wacker process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7758-89-6
Molecular Formula ClCu
Molecular Weight (g/mol) 98.996
MDL Number MFCD00010971
InChI Key OXBLHERUFWYNTN-UHFFFAOYSA-M
Synonym cuprous chloride, copper i chloride, copper chloride cucl, copper monochloride, dicopper dichloride, copper 1+ chloride, cucl, chlorid medny czech
PubChem CID 62652
ChEBI CHEBI:53472
IUPAC Name chlorocopper
SMILES Cl[Cu]

Specifications

Boiling Point 1490°C
Melting Point 430°C
Physical Form Powder
Quantity 500 g
Assay Percent Range 99% (Metal basis)
Solubility Information Very soluble in concentrated hydrochloric acid. Soluble in ammonium hydroxide. Insoluble in ethanol and acetone. Sparingly soluble in water.
Formula Weight 98.99
Refractive Index 1.93
Sensitivity Air and moisture sensitive
Appearance Powder
Density 4.14
Chemical Name or Material Copper(I) chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.