missing translation for 'onlineSavingsMsg'
Learn More

Copper(I) Cyanide, 0.99, Honeywell Fluka™

Product Code. 15672030 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Poly Bottle
Unit Size:
100g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15672030 5 g Poly Bottle 5g
15652030 100 g Poly Bottle 100g
15662030 500 g Poly Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15672030 Supplier Honeywell-Fluka Supplier No. 2163055G

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

0.99

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 544-92-3
Molecular Formula CCuN
Molecular Weight (g/mol) 89.564
MDL Number MFCD00010975
InChI Key DOBRDRYODQBAMW-UHFFFAOYSA-N
Synonym copper i cyanide, cupricin, copper +1 cyanide, copper cyanide cu cn, cyanocopper, rcra waste number p029, cucn, rcra waste no. p029, cyano copper
PubChem CID 11009
IUPAC Name copper(1+);cyanide
SMILES [C-]#N.[Cu+]

Specifications

Melting Point 473°C
Color Green or White to Yellow
Quantity 5 g
Linear Formula CuCN
UN Number UN1587
Beilstein 3587244
Formula Weight 89.56g/mol
Percent Purity 99%
Packaging Poly Bottle
Appearance Powder
Density 2.92g/mL at 25°C
Chemical Name or Material Copper(I) Cyanide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.