missing translation for 'onlineSavingsMsg'
Learn More

Copper(I) Iodide, Honeywell Fluka™

Product Code. 15691760 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Unit Size:
1kg
250g
50g
Product Code. Quantity unitSize
15691760 1 kg 1kg
15611770 50 g 50g
15601770 250 g 250g
3 options
This item is not returnable. View return policy

Product Code. 15691760

Brand: Honeywell-Fluka 2055401KG

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

0.98

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7681-65-4
Molecular Formula CuI
MDL Number MFCD00010978
Synonym cuprous iodide, copper l iodide, copper i iodide metals basis, marshite, copper i iodide 250g

Specifications

Quantity 1 kg
UN Number 3077
Formula Weight 190.45g/mol
Percent Purity 98%
Vapor Pressure 10mmHg (656°C)
Density 5.62g/mL at 25°C
Chemical Name or Material Copper(I) Iodide
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Copper(I) iodide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.