missing translation for 'onlineSavingsMsg'
Learn More

Copper(I) Oxide, Solstice

Product Code. 15611210 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
25 kg
6 x 2.5 kg
Unit Size:
2.5kg
25kg
500g
Pack of 6
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15611210 2.5 kg 2.5kg
15631210 500 g 500g
15641210 6 x 2.5 kg Pack of 6
15621210 25 kg 25kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15611210 Supplier Honeywell-Fluka Supplier No. 128412.5KG
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

≥86% Cu basis, red

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1317-39-1
Molecular Formula Cu2O
MDL Number MFCD00010974

Specifications

Color Red
Quantity 2.5 kg
UN Number 3077
Formula Weight 143.09g/mol
Percent Purity ≥86% (Cu basis)
Packaging Plastic Bottle
Chemical Name or Material Copper(I) Oxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.