Learn More
Copper(I) selenide, 99.5% (metals basis)
Description
Copper(I) selenide is used as intermediate in chemical research. It is a thermoelectric material used in a number of industrial applications including infrared detection and imaging.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 1113°C |
| Color | Black to Blue |
| Physical Form | Pieces |
| Quantity | 50 g |
| UN Number | UN3077 |
| Merck Index | 14,2666 |
| Solubility Information | Soluble in hydrochloric acid. Insoluble in water. |
| Formula Weight | 206.05 |
| Percent Purity | 99.5% |
| Density | 6.84 g/mL |
| Show More |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.