missing translation for 'onlineSavingsMsg'
Learn More

Copper(I) sulfide, 99.5% (metals basis)

Product Code. 11316489
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Unit Size:
1kg
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11316489 50 g 50g
11326489 250 g 250g
11306489 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11316489 Supplier Thermo Scientific Alfa Aesar Supplier No. 014718.18
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

For Methods: Luminous Paints, Electrodes, Solar Cells | CAS: 22205-45-4 | Cu2S | 159.15 g/mol

Copper(I) sulfide is used in luminous paints, electrodes and in solar cells. It is also used for grain mounts, XRD and in microprobe standards for the identification of unknown minerals. It acts as a precursor for the preparation of copper(I) oxide and copper metal. Further, it acts as a catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22205-45-4
Molecular Formula Cu2S
Molecular Weight (g/mol) 159.15
MDL Number MFCD00016062
InChI Key AQMRBJNRFUQADD-UHFFFAOYSA-N
Synonym copper i sulfide, cuprasulfide, copper sulfide cu2s, dicopper sulfide, dicopper sulphide, dicopper monosulfide, cuprous sulfide cu2s, unii-349m3c1rs1, cu2s, cupriosulfanyl copper
PubChem CID 62755
IUPAC Name bis(λ¹-copper(1+)) sulfanediide
SMILES [S--].[Cu+].[Cu+]

Specifications

Melting Point ∽1100°C
Color Black
Physical Form Powder
Quantity 50 g
Assay Percent Range 99.5% (Metal basis)
Solubility Information Slightly soluble in ammonium hydroxide. Insoluble in water and acetic acid.
Formula Weight 159.14
Odor Odorless
Appearance Black powder
Density 5.6 g/cm3
Chemical Name or Material Copper(I) sulfide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.