missing translation for 'onlineSavingsMsg'
Learn More

Copper(I) thiocyanate, 96% min

Product Code. 11339377
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
This item is not returnable. View return policy

Product Code. 11339377

Brand: Thermo Scientific Alfa Aesar 040220.30

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1111-67-7 | CHCuNS | 122.632 g/mol

It is used as flame retardant. It is a good inorganic pigment, and is used as the antifouling paint for protecting the underwater surfaces of ships against vegetation. Manufacturing industrial chemicals, pharmaceuticals and also used in freezing solutions, electroplating , steel picking, printing, and corrosion inhibitor against acid gases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1111-67-7
Molecular Formula CHCuNS
Molecular Weight (g/mol) 122.632
MDL Number MFCD00010980
InChI Key WEKJRGBVEBCTRF-UHFFFAOYSA-N
Synonym copper i thio-cyanate, thiocyanate ion copper
PubChem CID 16211891
IUPAC Name copper;thiocyanic acid
SMILES C(#N)S.[Cu]

Specifications

Melting Point 1084°C
Color White
Physical Form Powder
Quantity 250 g
Assay Percent Range 96% min.
Solubility Information Practically insoluble in water,alcohol. Soluble in NH4OH,ether
Formula Weight 121.63
Odor Odorless
Density 2.84
Chemical Name or Material Copper(I) thiocyanate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.