missing translation for 'onlineSavingsMsg'
Learn More

Copper(I) thiophene-2-carboxylate

Product Code. 15492379
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15492379 5 g 5g
15482379 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15492379 Supplier Thermo Scientific Alfa Aesar Supplier No. H64950.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 68986-76-5 | C5H3CuO2S | 190.683 g/mol

Reactant or reagent used for studies of xenobiotic response safener derivatives, synthesis of functionalized BODIPY dye analogs, orthogonal cross-coupling reactions, preparation of parent borondipyrromethene system and Copper mediated cross-coupling.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reactant or reagent used for studies of xenobiotic response safener derivatives, synthesis of functionalized BODIPY dye analogs, orthogonal cross-coupling reactions, preparation of parent borondipyrromethene system and Copper mediated cross-coupling.

Notes
Air and Light Sensitive. Protect from light store in dark. Store away from air. Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68986-76-5
Molecular Formula C5H3CuO2S
Molecular Weight (g/mol) 190.683
MDL Number MFCD02183524
InChI Key SFJMFSWCBVEHBA-UHFFFAOYSA-M
Synonym copper i thiophene-2-carboxylate, cutc, unii-7519wbl07l, copper i thiophenecarboxylate, copper 2-thienylcarboxylate, cuprous 2-thienylcarboxylate, copper 2-thiophenecarboxylate, copper 1+ thiophene-2-carboxylate, copper i thiophene-2-carboxylate mi
PubChem CID 11194830
IUPAC Name copper(1+);thiophene-2-carboxylate
SMILES C1=CSC(=C1)C(=O)[O-].[Cu+]

Specifications

Color Brown to Red
Physical Form Powder
Quantity 5 g
Formula Weight 190.68
Sensitivity Air and light sensitive
Chemical Name or Material Copper(I) thiophene-2-carboxylate
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Copper(I) thiophene-2-carboxylate
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Copper(I) thiophene-2-carboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.