missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) acetate, 99%, pure, anhydrous

Product Code. 10655671 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Plastic bottle
Unit Size:
1kg
250g
50g
This item is not returnable. View return policy

Product Code. 10655671

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 142-71-2 | C4H6CuO4 | 181.64 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142-71-2
Molecular Formula C4H6CuO4
Molecular Weight (g/mol) 181.64
InChI Key OPQARKPSCNTWTJ-UHFFFAOYSA-L
Synonym copper ii acetate, cupric acetate, copper diacetate, copper acetate, venus copper, copper 2+ diacetate, cupric diacetate, neutral verdigris, crystals of venus
PubChem CID 8895
IUPAC Name copper;diacetate
SMILES CC(=O)[O-].CC(=O)[O-].[Cu+2]

Specifications

Melting Point 115.0°C
Color Blue to Green
Physical Form Crystalline Powder
Quantity 50 g
Assay Percent Range 99%
Linear Formula (CH3CO2)2Cu
Fieser 01,159; 02,84; 03,65; 04,105; 05,156; 06,138; 10,103; 12,140; 15,99
Merck Index 15,2614
Solubility Information Solubility in water: soluble
Formula Weight 181.64
Percent Purity 99%
Grade Pure
Infrared Spectrum Authentic
Packaging Plastic bottle
Chemical Name or Material Copper(II) acetate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Copper(II) acetate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.