missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper(II) acetate, 99%, pure, anhydrous
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Plastic bottle
Unit Size:
1kg
250g
50g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Oxidation reagent
Specifications
Specifications
| Melting Point | 115.0°C |
| Color | Blue to Green |
| Physical Form | Crystalline Powder |
| Quantity | 250 g |
| Assay Percent Range | 99% |
| Linear Formula | (CH3CO2)2Cu |
| Fieser | 01,159; 02,84; 03,65; 04,105; 05,156; 06,138; 10,103; 12,140; 15,99 |
| Merck Index | 15,2614 |
| Solubility Information | Solubility in water: soluble |
| Formula Weight | 181.64 |
| Show More |
Safety and Handling
Product Identifier
- Copper(II) acetate
- Danger
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Acetic acid, copper(2+) salt
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction