missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper(II) acetate monohydrate, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 g
1000 g
5000 g
Description
Copper(II) acetate monohydrate is used in DNA extraction in biochemical applications. It is also used as a catalyst or oxidizing agent in organic syntheses.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Copper(II) acetate monohydrate is used in DNA extraction in biochemical applications. It is also used as a catalyst or oxidizing agent in organic syntheses.
Solubility
Soluble in water and alcohol. Slightly soluble in ether and glycerine.
Notes
Incompatible with strong oxidizing agents.
Copper(II) acetate monohydrate is used in DNA extraction in biochemical applications. It is also used as a catalyst or oxidizing agent in organic syntheses.
Solubility
Soluble in water and alcohol. Slightly soluble in ether and glycerine.
Notes
Incompatible with strong oxidizing agents.
Specifications
Melting Point | 115°C |
Quantity | 250 g |
Linear Formula | Cu(OOCCH3)2·H2O |
UN Number | UN3077 |
Merck Index | 14,2624 |
Solubility Information | Soluble in water and alcohol. Slightly soluble in ether and glycerine. |
Formula Weight | 199.65 (181.64 Anhydrous) |
Percent Purity | ≥98% |
Density | 1.882 g/mL |
Chemical Name or Material | Copper(II) acetate monohydrate |
Safety and Handling
Product Identifier
- Copper(II) acetate monohydrate
- Danger
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Copper diacetate monohydrate, Acetic acid, copper(2+) salt
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'