Learn More
Copper(II) acetate monohydrate, 99.9% (metals basis)
Description
Copper(II) acetate monohydrate is used in formation of Cu chelates of β-dicarbonyl compounds as a means of purification and in the eglinton (modified glaser) coupling of terminal alkynes. It is also used as a catalyst for the michael reaction giving increased yields and absence of side-reactions. It Promotes ullmann-type C-O and C-N coupling reactions of arylboronic acids with phenols, amines and various other nitrogen derivatives. It is also used in biochemical applications such as DNA extraction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 115°C |
| Color | Blue-Green |
| pH | 5.2 to 5.5 |
| Quantity | 5 g |
| Linear Formula | Cu(OOCCH3)2·H2O |
| UN Number | UN3077 |
| Merck Index | 14,2624 |
| Solubility Information | Soluble in water (72mg/ml at 20°C),alcohol,ether (slightly),and glycerin (slightly). |
| Formula Weight | 199.65 (181.64 Anhydrous) |
| Percent Purity | 99.9% |
| Show More |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.