missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper (II) Acetate Monohydrate, TRC
Click to view available options
Quantity:
50 g
Unit Size:
50g
Tekniske data
Tekniske data
| Chemical Name or Material | Copper (II) Acetate Monohydrate |
| CAS | 6046-93-1 |
| Molecular Formula | 2 C2 H3 O2 . Cu . H2 O |
| Quantity | 50 g |
| Synonym | Copper Diacetate Monohydrate, Copper(2+) Acetate Monohydrate, Copper(2+) Diacetate Monohydrate, Copper(II) Acetate Monohydrate, Cupric Acetate Monohydrate |
| InChI Formula | InChI=1S/2C2H4O2.Cu.H2O/c2*1-2(3)4;;/h2*1H3,(H,3,4);;1H2/q;;+2;/p-2 |
| SMILES | O.[Cu+2].CC(=O)[O-].CC(=O)[O-] |
| IUPAC Name | copper;diacetate;hydrate |
| Molecular Weight (g/mol) | 199.65 |
| Formula Weight | 198.9668 |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion