missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) Acetylacetonate, TRC

Product Code. 30356021
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30356021 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30356021 Supplier Toronto Research Chemicals Supplier No. TRCC685433250MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Copper(II) Acetylacetonate
CAS 13395-16-9
Molecular Formula C10H14CuO4
Quantity 250 mg
Synonym Copper, bis(2,4-pentanedionato-κO2,κO4)-, (SP-4-1)-, Copper, bis(2,4-pentanedionato)- (6CI,8CI), Copper, bis(2,4-pentanedionato-O,O')-, (SP-4-1)-, Copper, bis(2,4-pentanedionato-κO,κO')-, (SP-4-1)- (9CI), (SP-4-1)-Bis(2,4-pentanedionato-κO2,κO4)copper, Bis(2,4-pentanedionato)copper, Bis(acetylacetonato)copper, Bis(acetylacetone)copper, CD 9, Copper acetylacetonate, Copper bis(2,4-pentanedionate), Copper bis(acetylacetonate), Copper bis(acetylacetone), Copper diacetylacetonate, Copper(2+) bis(acetylacetonate), Copper(II) acetylacetonate, Copper(II) bis(2,4-pentanedionate), Cupric acetylacetonate, Cupric bis(2,4-pentanedionate), Nacem Copper
InChI Formula InChI=1S/2C5H8O2.Cu/c2*1-4(6)3-5(2)7;/h2*3,6H,1-2H3;/q;;+2/p-2
SMILES CC1=O[Cu+2]2(O=C(C)C=C(C)[O-]2)[O-]C(=C1)C
IUPAC Name copper;(Z)-4-oxopent-2-en-2-olate
Molecular Weight (g/mol) 261.76
Formula Weight 261.0188
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.