missing translation for 'onlineSavingsMsg'
Learn More

Copper(Ii) Carbonate Basic, Reagent grade, Honeywell™

Product Code. p-7120347 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
25 g
500 g
Packaging:
Poly Bottle
Unit Size:
25g
500g
This item is not returnable. View return policy

Product Code. 15641800

Brand: Honeywell Research Chemicals 20789625G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Reagent Grade

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12069-69-1
MDL Number MFCD00010976

Specifications

Melting Point 270°C
Color Blue to Green
Quantity 25 g
Linear Formula CuCO3 · Cu(OH)2
UN Number NONH for all modes of transport
Formula Weight 221.12g/mol
Grade Reagent
Packaging Poly Bottle
Appearance Powder
Density 4g/cu.cm
Chemical Name or Material Copper(Ii) Carbonate Basic
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.