missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) Carbonate, Technical, Basic, Fisher Chemical™

Product Code. 11423723 Shop All Fisher Chemical Products
missing translation for 'orderingAttributeHoverText'
Quantity:
500 g
3 kg
missing translation for 'unitSize'
3kg
500g
This item is not returnable. View return policy

Product Code. 11423723

missing translation for 'mfr': Fisher Chemical C/7879/53

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Copper(II) Carbonate, Technical, Basic

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12069-69-1
Molecular Formula CH2Cu2O5
Molecular Weight (g/mol) 221.11
MDL Number 10976
InChI Key ZMMDPCMYTCRWFF-UHFFFAOYSA-J
IUPAC Name dicopper(2+) dihydroxide carbonate
SMILES [OH-].[OH-].[Cu++].[Cu++].[O-]C([O-])=O

Specifications

Melting Point 200°C
Color Green
pH 8 to 9
Physical Form Powder/Solid
Quantity 500 g
Formula Weight 221.1g/mol
Packaging Plastic powder jar
compliance-icons
Product Identifier
  • Copper (II) carbonate basic
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.