missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) chloride, 99%, extra pure, anhydrous

Product Code. 10723724 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Plastic bottle
Unit Size:
1kg
2.5kg
250g
This item is not returnable. View return policy

Product Code. 10723724

Brand: Thermo Scientific Acros 206530025

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7447-39-4 | Cl2Cu | 134.45 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7447-39-4
Molecular Formula Cl2Cu
Molecular Weight (g/mol) 134.45
MDL Number MFCD00010972

Specifications

Boiling Point 993.0°C
Melting Point 498.0°C
Color Brown
Physical Form Fine Crystalline Powder
Quantity 2.5 kg
Assay Percent Range 98.5% min. (CuCl2)
Linear Formula CuCl2
Fieser 01,163; 02,84; 03,66; 04,105; 05,158; 06,139; 07,79; 08,119; 09,123; 10,106; 11,267; 13,152; 14,100
Merck Index 15, 2623
Synonym Cupric chloride
Solubility Information Solubility in water: 620g/L (20°C). Other solubilities: soluble in alcohol,acetone and ether
Formula Weight 134.45
Percent Purity 99%
Grade Extra Pure
Packaging Plastic bottle
Chemical Name or Material Copper(II) chloride, anhydrous
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Copper(II) chloride, anhydrous
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : copper chloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.