missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) Chloride Dihydrate, TRC

Product Code. 30356921
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30356921 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30356921 Supplier Toronto Research Chemicals Supplier No. TRCC685390250G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 10125-13-0
Quantity 250 g
Molecular Formula Cl2Cu . 2H2O
Synonym Copper Chloride, Copper Chloride Dihydrate, Copper Dichloride Dihydrate, Cupric Chloride Dihydrate
InChI Formula InChI=1S/2ClH.Cu.2H2O/h2*1H;;2*1H2/q;;+2;;/p-2
SMILES O.O.[Cl-].[Cl-].[Cu+2]
IUPAC Name dichlorocopper;dihydrate
Molecular Weight (g/mol) 170.48
Chemical Name or Material Copper(II) Chloride Dihydrate
Product Title
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.