missing translation for 'onlineSavingsMsg'
Learn More

Copper(II)-ethylenediamine complex, 1M solution in water

Product Code. p-93871 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
Unit Size:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10297060 1 L Glass bottle 1 L
11955191 2.5 L Glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 10297060 Proveedor Thermo Scientific Acros N.º de proveedor 258550010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Este artículo no se puede devolver. Vea la política de devoluciones

CAS: 14552-35-3 | C4H18CuN4O2 | 217.76 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Especificaciones

Chemical Name or Material Copper(II)-ethylenediamine complex
Name Note 1M Solution in Water
CAS 14552-35-3, 7732-18-5
Quantity 1 L
Molecular Formula C4H18CuN4O2
Linear Formula Cu(H2NCH2CH2NH2)2(OH)2
MDL Number MFCD00274628
Flash Point >110°C
Synonym Bis(ethylenediamine)copper(II)hydroxide
Solubility Information Solubility in water: soluble
Molecular Weight (g/mol) 217.76
Formula Weight 217.76
Packaging Glass bottle
Specific Gravity 1.1
Density 1.1000g/mL
Mostrar más Mostrar menos
compliance-icons
Nombre del producto
  • Copper(II)-ethylenediamine complex, 1M solution in water
Palabra de advertencia
  • Danger
Clasificación
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Indicaciones de peligro
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Consejos de prudencia
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.