Learn More
Copper(II) gluconate
Description
Copper(II) gluconate is used in dietary supplement. It is also used to treat acne vulgaris, common cold, hypertension, premature labor, leishmaniasis and visceral postoperative complications. Further, it is used in oral deodorants and feed additives. In addition to this, it is used as a synergist and nutritional supplement for foods. It is the active component in retsyn used in mints and certs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 155°C to 157°C |
| Quantity | 1 kg |
| Merck Index | 14,2640 |
| Solubility Information | Soluble in water. Insoluble in acetone,alcohol and ether. |
| Formula Weight | 453.85 |
| Physical Form | Powder |
| Chemical Name or Material | Copper(II) gluconate |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.