missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) hexafluoro-2,4-pentanedionate hydrate, 99.99% (metals basis)

Product Code. 11384847
Change view
Click to view available options
Quantity:
0.5 g
2 g
10 g
Unit Size:
0.5g
10g
2g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11384847 2 g 2g
11374847 0.5 g 0.5g
11364847 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11384847 Supplier Thermo Scientific Alfa Aesar Supplier No. 041870.04
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 155640-85-0 | C10H2CuF12O4 | 477.65 g/mol

Used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 155640-85-0
Molecular Formula C10H2CuF12O4
Molecular Weight (g/mol) 477.65
MDL Number MFCD00151019
InChI Key HZXGNBMOOYOYIS-PAMPIZDHSA-L
Synonym copper ii hexafluoroacetylacetonate hydrate, bis hexafluoroacetylacetonato copper ii hydrate, copper ii hexafluoro-2,4-pentanedionate hydrate, copper hexafluoroacetylacetonate hydrate, copper ii hexafluoroacetyl-acetonate hydrate, hexafluoroacetylacetonato copper ii hydrate, copper ii hexafluoroacetylacetonate hydrate, elec. gr., bis 1,1,1,5,5,5-hexafluoroacetylacetonato copper ii hydrate, copper ii 1,1,1,5,5,5-hexafluoro-2,4-pentanedionate hydrate, 3z-1,1,1,5,5,5-hexafluoro-4-2z-1,1,1,5,5,5-hexafluoro-4-oxopent-2-en-2-yl oxy cuprio oxy pent-3-en-2-one hydrate
PubChem CID 56845342
SMILES [Cu++].[O-]\C(=C/C(=O)C(F)(F)F)C(F)(F)F.[O-]\C(=C/C(=O)C(F)(F)F)C(F)(F)F

Specifications

Boiling Point 220°C (decomposition)
Melting Point ∼100°C
Physical Form Crystalline
Quantity 2 g
Solubility Information Soluble in methanol,acetone,toluene
IUPAC Name copper(2+) bis((2Z)-1,1,1,5,5,5-hexafluoro-4-oxopent-2-en-2-olate)
Formula Weight 477.64 (Anhydrous)
Percent Purity 99.99%
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Copper(II) hexafluoro-2,4-pentanedionate hydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.