missing translation for 'onlineSavingsMsg'
Learn More

Copper(Ii) Nitrate Hemi(Pentahydrate), 98.0 to 102.0%, ACS Reagent, Honeywell™ Fluka™

Product Code. 15662200 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Poly Bottle
Unit Size:
100g
2.5kg
500g
This item is not returnable. View return policy

Product Code. 15662200

Brand: Honeywell-Fluka 223395100G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

ACS Reagent, 98%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19004-19-4
MDL Number MFCD02253001
Synonym dicopper tetranitrate pentahydrate, copper ii nitrate hemipentahydrate, cupric nitrate, hydrate, copper, reference standard solution, copper nitrate hemipentahydrate, cupric nitrate hemipentahydrate, 2cu.4no3.5h2o, cupric nitrate hemi pentahydrate

Specifications

Quantity 100 g
Linear Formula Cu(NO3)2 · 2.5H2O
UN Number UN3085
Formula Weight 232.59g/mol
Percent Purity 98.0 to 102.0%
Grade ACS Reagent
Packaging Poly Bottle
Chemical Name or Material Copper(Ii) Nitrate Hemi(Pentahydrate)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.