missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) nitrate hemi(pentahydrate), 98%

Product Code. 11304549
Click to view available options
Quantity:
500 g
Unit Size:
500g
This item is not returnable. View return policy

Product Code. 11304549

Brand: Thermo Scientific Alfa Aesar A14004.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 19004-19-4 | Cu2H10N4O17 | 465.18 g/mol

Copper(II) nitrate hemi(pentahydrate) is used for the catalytic oxidative coupling of 2,6-dimethylphenol to produce a polymer which is an important engineering material. Its solutions are used in textiles and polishing agents for other metals. It is commonly used in school laboratories to demonstrate chemical voltaic cell reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Copper(II) nitrate hemi(pentahydrate) is used for the catalytic oxidative coupling of 2,6-dimethylphenol to produce a polymer which is an important engineering material. Its solutions are used in textiles and polishing agents for other metals. It is commonly used in school laboratories to demonstrate chemical voltaic cell reactions.

Solubility
Soluble in water, ammonia and ethanol

Notes
Incompatible with reducing agents, organic materials and powdered metals.
TRUSTED_SUSTAINABILITY

Specifikationer

Boiling Point 170°C (decomposition)
Melting Point ∼115°C
CAS 19004-19-4
Quantity 500 g
Molecular Formula Cu2H10N4O17
Linear Formula Cu(NO3)2·2.5 H2O
MDL Number MFCD00149671
UN Number UN3085
Merck Index 14,2643
Synonym dicopper tetranitrate pentahydrate, copper ii nitrate hemipentahydrate, cupric nitrate, hydrate, copper, reference standard solution, copper nitrate hemipentahydrate
Solubility Information Soluble aq. soln.,ammonia and ethanol
InChI Key IZDJJEMZQZQQQQ-UHFFFAOYSA-N
SMILES O.O.O.O.O.[Cu++].[Cu++].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
IUPAC Name dicopper(2+) pentahydrate tetranitrate
Molecular Weight (g/mol) 465.18
Formula Weight 232.59 (187.55 Anhydrous)
Percent Purity 98%
Odor Odorless
Sensitivity Hygroscopic
Density 2.32
Chemical Name or Material Copper(II) nitrate hemi(pentahydrate)
Visa mer Visa mindre

RUO – Research Use Only

Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.