missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) nitrate hydrate, Puratronic™, 99.999% (metals basis)

Product Code. 15415965
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Unit Size:
100g
25g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15415965 25 g 25g
11380959 5 g 5g
11300969 100 g 100g
11310969 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15415965 Supplier Thermo Scientific Alfa Aesar Supplier No. 010699.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13778-31-9 | CuN2O6 | 187.55 g/mol

It serves as a catalyst for oxidative coupling of 2,6-dimethylphenol, resulting in an important engineering polymer material. Also used in textiles and polishing agents for other metals. It is a precursor for copper oxide, which finds use as a catalyst for many organic transformations. Menke conditions employ copper nitrate for aromatic nitrations. Claycop reagent, the hydrated copper nitrate deposited on clay, finds use in many organic reactions, for example, oxidation of dithioacetals to carbonyls and thiols to disulfides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13778-31-9
Molecular Formula CuN2O6
Molecular Weight (g/mol) 187.55
MDL Number MFCD00149669
InChI Key XTVVROIMIGLXTD-UHFFFAOYSA-N
Synonym copper ii nitrate hydrate, copper nitrate hydrate, acmc-20aldm, cupric nitrate hydrate, cu.2no3.h2o, copper 2+ ion hydrate dinitrate, copper 2+ hydrate dinitrate, copper ii nitrate hydrate, puratronic, copper 2+ nitrate-water 1/2/1, nitric acid, copper 2+ salt, hydrate 8ci,9ci
PubChem CID 15303652
SMILES [Cu++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Color Blue
Physical Form Crystalline Aggregates
Quantity 25 g
Assay Percent Range 99.999% (Metals basis)
Solubility Information Soluble in ethanol and water. Insoluble in ethyl acetate.
IUPAC Name copper;dinitrate;hydrate
Formula Weight 187.55 (anhydrous)
Odor Odorless
Sensitivity Hygroscopic
Chemical Name or Material Copper(II) nitrate hydrate
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Copper(II) nitrate hydrate
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Oxidising solid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P371+P380+P375-In case of major fire and large quantities: Evacuate area Fight fire remotely due to the risk of explosion.
Supplemental information
  • MIXTURE LIST-Contain: Copper(II) nitrate hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.