missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) nitrate trihydrate, Solstice

Product Code. 15606030
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15606030 500 g 500g
15696020 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15606030 Supplier Honeywell-Fluka Supplier No. 61194500G
  • ONLINE EXCLUSIVE PRICE
    214.00 €
    192.83 € / 500g
    Save 21.17 €
    9% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Puriss. p.a., 99-104%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10031-43-3
Molecular Formula CuH6N2O9
Molecular Weight (g/mol) 241.599
MDL Number MFCD00149671
InChI Key SXTLQDJHRPXDSB-UHFFFAOYSA-N
Synonym copper ii nitrate trihydrate, copper nitrate trihydrate, cupric nitrate trihydrate, copper ii nitrate hydrate, copper ii nitrate 3-hydrate, copper 2+ trihydrate dinitronate, copper 2+ trihydrate dinitrate, copper ii nitrate trihydrate-cu puratrem
PubChem CID 9837674
IUPAC Name copper;dinitrate;trihydrate
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.[Cu+2]

Specifications

Physical Form Powder
Quantity 500 g
Linear Formula Cu(NO3)2 · 3H2O
UN Number UN1477
Formula Weight 241.6g/mol
Percent Purity 99 to 104%
Chemical Name or Material Copper(II) nitrate trihydrate
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Copper(II) nitrate trihydrate
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Substances and mixtures corrosive to metals Category 1
  • Oxidising solid Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H271-May cause fire or explosion; strong oxidiser.
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P234-Keep only in original container.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Copper(II) nitrate trihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.