missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Leading the Industry For Over 65 Years
- Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Quantity | 100 g |
| Linear Formula | Cu(NO3)2 · 3H2O |
| UN Number | UN1477 |
| Formula Weight | 241.6g/mol |
| Percent Purity | 99 to 104% |
| Chemical Name or Material | Copper(II) nitrate trihydrate |
Safety and Handling
Product Identifier
- Copper(II) nitrate trihydrate
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Substances and mixtures corrosive to metals Category 1
- Oxidising solid Category 1
- Skin corrosion/irritation Category 1 B
- H271-May cause fire or explosion; strong oxidiser.
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P234-Keep only in original container.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-[In case of inadequate ventilation] wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contain: Copper(II) nitrate trihydrate
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?