missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) Oxide, ≥99% (RT), Granular, Honeywell™ Fluka™

Product Code. p-7120352 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
50 g
250 g
Packaging:
Poly Bottle
Unit Size:
250g
50g
This item is not returnable. View return policy

Product Code. 15637540

Brand: Honeywell-Fluka 9473750G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

≥99.0% (RT), granular

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1317-38-0
Molecular Formula CuO
Molecular Weight (g/mol) 79.55
MDL Number MFCD00010979
InChI Key KKCXRELNMOYFLS-UHFFFAOYSA-N
Synonym cupric oxide, copper ii oxide, copper oxide, copper monoxide, banacobru ol, chrome brown, copper brown, copper monooxide, black copper oxide
PubChem CID 14829
SMILES [O--].[Cu++]

Specifications

Melting Point 1336°C
Color Black
Physical Form Powder
Quantity 50 g
UN Number UN3077
IUPAC Name oxocopper
Formula Weight 79.55g/mol
Percent Purity ≥99% (RT)
Odor Odorless
Packaging Poly Bottle
Density 6.450g/cm3 at 20°C
Chemical Name or Material Copper(II) oxide
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.