missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) oxide, nanopowder

Product Code. 11489850
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11489850 50 g 50g
11479850 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11489850 Supplier Thermo Scientific Alfa Aesar Supplier No. 044663.18
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1317-38-0 | CuO | 79.55 g/mol

Copper(II) oxide nanopowder is used as a burning rate catalyst in rocket propellant, which enhances the the homogeneous propellant burning rate, lower pressure index. It can act as a catalyst used in superconducting materials, thermoelectric materials, sensing materials, glass and ceramics. It plays a vital role in photoconductive and photothermal applications. It is also used in ceramic resistors, magnetic storage media, gas sensors, near-infrared tilters, semiconductors, solar energy transformation and high-tech superconductors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1317-38-0
Molecular Formula CuO
Molecular Weight (g/mol) 79.55
MDL Number MFCD00010979
InChI Key KKCXRELNMOYFLS-UHFFFAOYSA-N
Synonym cupric oxide, copper ii oxide, copper oxide, copper monoxide, banacobru ol, chrome brown, copper brown, copper monooxide, black copper oxide
PubChem CID 14829
SMILES [O--].[Cu++]

Specifications

Melting Point 1326°C
Color Black
Physical Form Powder
Quantity 50 g
UN Number UN3077
Merck Index 14,2646
Solubility Information Practically insoluble in water,and alcohol. Soluble in dilute acids,alkali cyanides,and ammonium carbonate solution.
IUPAC Name oxocopper
Formula Weight 79.55
Odor Odorless
Refractive Index 2.63
Density 6.3 g/mL
Chemical Name or Material Copper (II) oxide
Show More Show Less
compliance-icons
Product Identifier
  • Copper(II) oxide, nanopowder
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P391-Collect spillage.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Copper oxide (CuO)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.