missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) phosphate, 98%

Product Code. 11335079
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11335079 100 g 100g
11345079 500 g 500g
11325079 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11335079 Supplier Thermo Scientific Alfa Aesar Supplier No. A17360.22
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7798-23-4 | Cu3O8P2 | 380.578 g/mol

Copper(II) phosphate is used as organic catalyst, fertilizer, emulsifier, corrosion inhibitor for phosphoric acid, metal protectant, and animal feed additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Copper(II) phosphate is used as organic catalyst, fertilizer, emulsifier, corrosion inhibitor for phosphoric acid, metal protectant, and animal feed additive.

Solubility
Insoluble with water and ethanol. Soluble in ammonia, ammonium hydroxide, acetone.

Notes
Hygroscopic. Incompatible with strong bases, metals, alkali metals and powdered metals.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7798-23-4
Molecular Formula Cu3O8P2
Molecular Weight (g/mol) 380.578
MDL Number MFCD00036268
InChI Key GQDHEYWVLBJKBA-UHFFFAOYSA-H
Synonym copper ii phosphate, cupric phosphate, unii-n8np6fr80r, tricopper bis orthophosphate, n8np6fr80r, copper phosphate cu3 po4 2, copper phosphate 3:2, hsdb 267, cupric phosphate cu3 po4 2, phosphoric acid, copper 2+ salt
PubChem CID 86469
IUPAC Name tricopper;diphosphate
SMILES [O-]P(=O)([O-])[O-].[O-]P(=O)([O-])[O-].[Cu+2].[Cu+2].[Cu+2]

Specifications

Melting Point >300°C
Quantity 100 g
Linear Formula Cu3(PO4)2
Merck Index 14,2648
Solubility Information Insoluble with water and ethanol. Soluble in ammonia,ammonium hydroxide,acetone.
Formula Weight 380.58
Percent Purity 98%
Odor Odorless
Chemical Name or Material Copper(II) phosphate
compliance-icons compliance-icons
Product Identifier
  • Copper(II) phosphate
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Copper(II) phosphate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.